Learn More
Worthington Biochemical Corporation COLLAGNASE 250MG (5 PK 50MG)
SDP
Catalog No. NC9153526
Encompass
Collagenase, Type 3, Filtered, which is filtered through a 0.22 Micron membrane and lyophilized in vials to contain > /=50mg/VL, Store at 2 to 8degreeC, Pack Size: 5 x 50mg, Source: Clostridium histolyticum, Minimum Activity: > /=100U/mg dr
Catalog No. NC9153526 Supplier Worthington Biochemical Corporation Supplier No. LS004208
Collagenase, Type 3, Filtered, which is filtered through a 0.22 Micron membrane and lyophilized in vials to contain > /=50mg/VL, Store at 2 to 8degreeC, Pack Size: 5 x 50mg, Source: Clostridium histolyticum, Minimum Activity: > /=100U/mg dr