Learn More
Worthington Biochemical Corporation Collagenase, Type 3, Filtered, 50mg
SDP
Catalog No. NC9027439
Encompass
Collagenase, Type 3 (Code: CLS-3), which is filtered through a 0.22 micron membrane and lyophilized in vials to contain 50 milligrams per vial. Store at 2-8°C.
Catalog No. NC9027439 Supplier Worthington Biochemical Corporation Supplier No. LS004206
Collagenase, Type 3 (Code: CLS-3), which is filtered through a 0.22 micron membrane and lyophilized in vials to contain 50 milligrams per vial. Store at 2-8°C.