Learn More
Med Vet International KVP Bird Collar, 10in. Shield Diameter, 4.7in. - 5.5in. Neck Opening
Catalog No. 502830655
Encompass_Preferred
KVP 10in. Saf-T Shield collar for birds has a 4.7in. – 5.5in. neck opening circumference, an ultra-light construction, and an exclusive dual padded neckline. This collar is made of a clear lexan that will not impair peripheral vision.
Catalog No. 50-283-0655 Supplier Med Vet International Supplier No. EJ410X
KVP 10in. Saf-T Shield collar for birds has a 4.7in. – 5.5in. neck opening circumference, an ultra-light construction, and an exclusive dual padded neckline. This collar is made of a clear lexan that will not impair peripheral vision.