Learn More
Med Vet International 3M Collar, Ft-15, Each
Catalog No. 502830776
Encompass_Preferred
This fit test collar is compatible with a selection of test kits for ease of use It tests the respirator seal to make sure it's working properly This 3M™ collar is virtually universally compatible and works with most gas and vapor and particulate respirators It conforms to OSHA regulations to keep you safe This respirator collar is part of a full test environment for use with a bitter or sweet solution
Catalog No. 50-283-0776 Supplier Med Vet International Supplier No. 231443EA
This fit test collar is compatible with a selection of test kits for ease of use It tests the respirator seal to make sure it's working properly This 3M™ collar is virtually universally compatible and works with most gas and vapor and particulate respirators It conforms to OSHA regulations to keep you safe This respirator collar is part of a full test environment for use with a bitter or sweet solution