Learn More
Ace Glass, Inc. Collar, glass filled PTFE, 28mm, w/PTFE gasket
SDP
Catalog No. 50976106
Encompass_Preferred
28mm glass filled collar to be used with ACE stir shafts. Supplied with PTFE gasket to prevent scratching top of bearing and acts as dust cover.
Catalog No. 50-976-106 Supplier Ace Glass, Inc. Supplier No. 812728
28mm glass filled collar to be used with ACE stir shafts. Supplied with PTFE gasket to prevent scratching top of bearing and acts as dust cover.