Learn More
Med Vet International Jorvet Soft E-Collar, Large, J1003G
Catalog No. 5000495780
Encompass_Preferred
The Jorvet large soft e-collar was designed for the comfort of the patient and is still effective in offering licking and chewing protection for animals. This elizabethan type collar allows pets to move about freely without damaging furniture, and they are able to eat and drink more freely. The soft E-Collar is made of a heavy nylon materials that is waterproof and held in place by a strong flat tie.
Catalog No. 50-004-95780 Supplier Med Vet International Supplier No. JOR1003G
The Jorvet large soft e-collar was designed for the comfort of the patient and is still effective in offering licking and chewing protection for animals. This elizabethan type collar allows pets to move about freely without damaging furniture, and they are able to eat and drink more freely. The soft E-Collar is made of a heavy nylon materials that is waterproof and held in place by a strong flat tie.