Learn More
Med Vet International Jorvet Soft E-Collar, SS/Medium, For Cats/Dogs 15-20lbs, J1003D
Catalog No. 5000495730
Encompass_Preferred
The Jorvet medium soft e-collar was designed for the comfort of the patient and is still effective in offering licking and chewing protection for animals. This elizabethan type collar allows pets to move about freely without damaging furniture, and they are able to eat and drink more freely. The soft E-Collar is made of a heavy nylon materials that is waterproof and held in place by a strong flat tie.
Catalog No. 50-004-95730 Supplier Med Vet International Supplier No. JOR1003D
The Jorvet medium soft e-collar was designed for the comfort of the patient and is still effective in offering licking and chewing protection for animals. This elizabethan type collar allows pets to move about freely without damaging furniture, and they are able to eat and drink more freely. The soft E-Collar is made of a heavy nylon materials that is waterproof and held in place by a strong flat tie.