Learn More
Ace Glass, Inc. Collar, stainless steel, 19mm, w/PTFE gasket
SDP
Catalog No. 50976107
Encompass_Preferred
19mm 316 stainless steel collar to be used with ACE stir shafts. Supplied with PTFE gasket to prevent scratching top of bearing and acts as dust cover.
Catalog No. 50-976-107 Supplier Ace Glass, Inc. Supplier No. 812743
19mm 316 stainless steel collar to be used with ACE stir shafts. Supplied with PTFE gasket to prevent scratching top of bearing and acts as dust cover.