Learn More
Med Vet International Thyroid X-Ray Collar, J0678N, Each
Catalog No. 5000495088
Encompass_Preferred
New style Thyroid Collar offers a trim yet shielded and comfortable design. Includes binding and rear hook and loop closure. 25.5” length, 10 x 5.5” Bib.• Standard protection: 0.5mm Pb Equivalency• Royal blue nylon(Please specify color combinations. This item is a special order and non-returnable.)
Catalog No. 50-004-95088 Supplier Med Vet International Supplier No. JOR678N
New style Thyroid Collar offers a trim yet shielded and comfortable design. Includes binding and rear hook and loop closure. 25.5” length, 10 x 5.5” Bib.• Standard protection: 0.5mm Pb Equivalency• Royal blue nylon(Please specify color combinations. This item is a special order and non-returnable.)