Learn More
GENTUERI INC CollectEject(tm) Oral Swab, Sterilized, Non-Medical or Class 1 general medical use
SDP
Catalog No. NC2146463
Encompass_Preferred
The multi-layered design maximizes the collection of samples since the layers separate when the swab becomes wet. This increases the surface area and allows for a larger surface area for collection and a more efficeint extraction of the sample.
This item is not returnable. View return policy
Catalog No. NC2146463 Supplier GENTUERI INC Supplier No. CE110S
The multi-layered design maximizes the collection of samples since the layers separate when the swab becomes wet. This increases the surface area and allows for a larger surface area for collection and a more efficeint extraction of the sample.