Learn More
GENTUERI INC CollectEject™ Touch DNA Swab
SDP
Catalog No. NC2146466
Encompass_Preferred
Touch DNA refers to DNA that can be obtained from skin cells left behind at crime scenes or surfaces. The small amount of sample that is available must be collected carefully to maximize what is collected and extracted without introducing contamination. The small design of the CollectEject™ Touch DNA swab head helps to concentrate the sample and minimizes the amount of extraction buffer required for processing. This design increases the chances of generating a successful DNA profile with the fewest manipulations, reducing the potential of introducing contaminates.Key Features•Small head designed to concentrate samples from fingerprints•Minimizes the amount of extraction buffer required for processing•Ejecting the head reduces manipulations and chances of introducing contamination
This item is not returnable. View return policy
Catalog No. NC2146466 Supplier GENTUERI INC Supplier No. CE120S
Touch DNA refers to DNA that can be obtained from skin cells left behind at crime scenes or surfaces. The small amount of sample that is available must be collected carefully to maximize what is collected and extracted without introducing contamination. The small design of the CollectEject™ Touch DNA swab head helps to concentrate the sample and minimizes the amount of extraction buffer required for processing. This design increases the chances of generating a successful DNA profile with the fewest manipulations, reducing the potential of introducing contaminates.Key Features•Small head designed to concentrate samples from fingerprints•Minimizes the amount of extraction buffer required for processing•Ejecting the head reduces manipulations and chances of introducing contamination