Learn More
Electron Microscopy Sciences Disposable Collecting Bottles, 100-1000 ml;50 ml, Sterile
SDP
Catalog No. 5030920
Encompass_Preferred
Disposable polystyrene bottles for the storage of media, buffers and other aqueous solutions. Easy grip sides facilitate handling. Plug seal cap provides an airtight seal and helps minimize the risk of contamination. Sterile, certified non-pyrogenic. Neck size 45 mm.
Type: 100-1000 ml;50 ml
Sterile: Sterile
Catalog No. 50-309-20 Supplier Electron Microscopy Sciences Supplier No. 6424410
Disposable polystyrene bottles for the storage of media, buffers and other aqueous solutions. Easy grip sides facilitate handling. Plug seal cap provides an airtight seal and helps minimize the risk of contamination. Sterile, certified non-pyrogenic. Neck size 45 mm.
Type: 100-1000 ml;50 ml
Sterile: Sterile