Learn More
Fritsch Milling And Sizing Inc Collecting Vessel 10 Liters for P-19/P-25
SDP
Catalog No. 501534499
Encompass
Collecting vessel 10 litres for PULVERISETTE 19 and PULVERISETTE 25 for grinding large quantities. Collecting vessels are available in a range of sizes. An adapter is required when these are used in combination with sample extraction equipment using the cyclone.
Catalog No. 50-153-4499 Supplier Fritsch Milling And Sizing Inc Supplier No. 45.5700.00
Collecting vessel 10 litres for PULVERISETTE 19 and PULVERISETTE 25 for grinding large quantities. Collecting vessels are available in a range of sizes. An adapter is required when these are used in combination with sample extraction equipment using the cyclone.