Learn More
Fritsch Milling And Sizing Inc Collecting Vessel 3 Liters for P-19/P-25
SDP
Catalog No. 501534500
Encompass
Collecting vessels (stainless steel) for PULVERISETTE 19 and PULVERISETTE 25 are available in a range of sizes.
This collecting vessel has a volume of 3 litres.
For grinding large quantities the 10 Litre volume collecting vessel is recommended.
Catalog No. 50-153-4500 Supplier Fritsch Milling And Sizing Inc Supplier No. 45.5730.00
Collecting vessels (stainless steel) for PULVERISETTE 19 and PULVERISETTE 25 are available in a range of sizes.
This collecting vessel has a volume of 3 litres.
For grinding large quantities the 10 Litre volume collecting vessel is recommended.