Learn More
Fritsch Milling And Sizing Inc P-16 Collecting Vessel 30 Litres with Filter Hose
SDP
Catalog No. 501534015
Encompass
As standard the PULVERISETTE 16 is delivered with a collecting vessel 5 litres. For grinding large quantities this collecting vessel with a capacity of 30 litres and filter hose is recommended.
Catalog No. 50-153-4015 Supplier Fritsch Milling And Sizing Inc Supplier No. 16.6124.00
As standard the PULVERISETTE 16 is delivered with a collecting vessel 5 litres. For grinding large quantities this collecting vessel with a capacity of 30 litres and filter hose is recommended.