Learn More
Fritsch Milling And Sizing Inc Collecting Vessel 56 Litres Stainless Steel
SDP
Catalog No. 501534519
Encompass
Collecting vessel 56 litres stainless steel for high-performance Cyclone separator 19.1900.00
Is recommended, if a larger sample collection than 1000 ml is required, because the high-performance Cyclone separator is delivered with a 1000 ml glass bottle as standard. Also 2 litres and 5 litres of sample glasses are available.
Catalog No. 50-153-4519 Supplier Fritsch Milling And Sizing Inc Supplier No. 45.8050.00
Collecting vessel 56 litres stainless steel for high-performance Cyclone separator 19.1900.00
Is recommended, if a larger sample collection than 1000 ml is required, because the high-performance Cyclone separator is delivered with a 1000 ml glass bottle as standard. Also 2 litres and 5 litres of sample glasses are available.