Learn More
Med Vet International McKesson Consult™ Urine Transfer Straw, 3 Inch, 16-364966UT, 100/Box
Catalog No. 503043340
Encompass_Preferred
Urine Transfer Straw McKesson Consult™ 3 Inch (7.6 cm) Length: 3 inches (7.6 centimeters) Leak resistant Helps reduce spills, improves lab hygiene, and limit exposure to dangerous samples Helps reduce the risk of contamination for improved test results Transfers samples to a tube suitable for analysis and transport Compatible with most vacuum urine tubes Provides consistent sample transfer volumes when used with like vacuum tubes
Catalog No. 50-304-3340 Supplier Med Vet International Supplier No. 1194144BX
Urine Transfer Straw McKesson Consult™ 3 Inch (7.6 cm) Length: 3 inches (7.6 centimeters) Leak resistant Helps reduce spills, improves lab hygiene, and limit exposure to dangerous samples Helps reduce the risk of contamination for improved test results Transfers samples to a tube suitable for analysis and transport Compatible with most vacuum urine tubes Provides consistent sample transfer volumes when used with like vacuum tubes