Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Collection Flask/Beaker

Catalog No. 111010871 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Capacity (Metric):
300 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
11-101-0871 300 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-0871 Supplier MilliporeSigma™ Supelco™ Supplier No. 21266U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for use in the Food and Beverage Industry

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 300 mL
For Use With (Application) Food and Beverages
No. per Pack 3/Pk.
Product Type Collection Flask/Beaker
Bottom Shape Flat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.