Learn More
Med Vet International UTM Nasal Collection and Transport System Sterile, CEPH3C064N, 50/Box
Catalog No. 503044773
Encompass_Preferred
3 mL of UTM-RT medium in 16x100 mm screw-cap tube with internal shaped conical bottom One regular size applicator swab flocked tipped with breaking point UTM Universal Transport Medium Universal Transport Media without beads is an FDA cleared collection and transport system suitable for collection, transport, preservation, and long-term freeze storage of clinical specimens containing viruses for viral molecular diagnostic testing, and meets CDC Interim Guidelines for Collecting, Handling, and Testing Clinical COVID-19 Specimens
Catalog No. 50-304-4773 Supplier Med Vet International Supplier No. 1217792BX
3 mL of UTM-RT medium in 16x100 mm screw-cap tube with internal shaped conical bottom One regular size applicator swab flocked tipped with breaking point UTM Universal Transport Medium Universal Transport Media without beads is an FDA cleared collection and transport system suitable for collection, transport, preservation, and long-term freeze storage of clinical specimens containing viruses for viral molecular diagnostic testing, and meets CDC Interim Guidelines for Collecting, Handling, and Testing Clinical COVID-19 Specimens