Learn More
Waters Corp Waters Sample Collection Plate, 96-Well, 1 mL, Polypropylene
Catalog No. 50818605 Shop All Waters Corporation Products
Encompass_Preferred
This 96-well sample collection plate features 1 mL wells and is SBS/ANSI compliant for robot-compatible systems. It can also function as a collection plate for 96-well SPE and filtration-plate formats.
  • 1 mL volume capacity per well.
  • SBS/ANSI compliant for robot compatible systems.
  • Compatible with most automated liquid handling systems.
  • Made of chemically resistant material, ensuring compatibility with a wide range of solvents and reagents.
  • Designed for long-term sample storage, providing excellent sample integrity and minimizing evaporation.
  • Pyramid-shaped bottom.
Catalog No. 50-818-605 Supplier Waters Corp Supplier No. WAT058957
This 96-well sample collection plate features 1 mL wells and is SBS/ANSI compliant for robot-compatible systems. It can also function as a collection plate for 96-well SPE and filtration-plate formats.
  • 1 mL volume capacity per well.
  • SBS/ANSI compliant for robot compatible systems.
  • Compatible with most automated liquid handling systems.
  • Made of chemically resistant material, ensuring compatibility with a wide range of solvents and reagents.
  • Designed for long-term sample storage, providing excellent sample integrity and minimizing evaporation.
  • Pyramid-shaped bottom.