Learn More
Zymo Research Corporation Collection Plate, 2 plates
SDP
Catalog No. 50444078
Encompass_Preferred
Each has a level footprint and conforms to laboratory standards. Adaptable for use with either Silicon-A, Zymo-Spin I-96, Zymo-Spin™ IB-96, and Zymo-Spin III-96 plates. Capacity is 1.2 ml per round bottom well.
Catalog No. 50-444-078 Supplier Zymo Research Corporation Supplier No. C2002
Each has a level footprint and conforms to laboratory standards. Adaptable for use with either Silicon-A, Zymo-Spin I-96, Zymo-Spin™ IB-96, and Zymo-Spin III-96 plates. Capacity is 1.2 ml per round bottom well.