Learn More
Waters Corp WATERS CORPORATION Collection Rack, Eppendorf Tubes, 120 Capacity
Catalog No. 50793109 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This collection rack is designed for use with the Waters Fraction Collector III. It provides a convenient solution for collecting purified peak fractions into Eppendorf tubes.
  • Accepts up to 120 eppendorf tubes.
  • Compatible with the Fraction Collector III.
  • Suitable for collecting purified peak fractions.
Catalog No. 50-793-109 Supplier Waters Corp Supplier No. 725000145
This collection rack is designed for use with the Waters Fraction Collector III. It provides a convenient solution for collecting purified peak fractions into Eppendorf tubes.
  • Accepts up to 120 eppendorf tubes.
  • Compatible with the Fraction Collector III.
  • Suitable for collecting purified peak fractions.