Description
- Heat-sealed integrity pouch
- Designed for the collection and transport of urine samples
- Helps prevent subjects from using someone elses urine
- Features a concealed Temperature-monitoring thermometer beneath a Tear-strip label.
- Allows collectors to verify specimen temperature only after collection.
Specifications
Specifications
| Product Type | Urine Specimen Container |
| Disposable | Single-use |
| Graduated | No |
| Material | Plastic |
| Sterility | Non-sterile |
| Includes | Concealed Thermometer |
| No. per Pack | 100/Pk. |
| Volume (Metric) | 100 ml |