Learn More
Instech Collection tube, 500µL, K3EDTA, violet (Sarstedt Microvette 500 20.1341.102)
SDP
Catalog No. NC2580246
Encompass
Automated Blood Sampling Collection Tubes
The Microvette 300 and 500 tubes are ideal for use with the Instech Automated Blood Sampler due to the shape of the tube, which simplifies pipetting, and the wide range of additives and fill volumes.
The CT-M500-LIHEPG and CT-M500-SG models contain a unique inert acrylic gel at the bottom of the tube, under the additive, that moves upward during centrifugation to form a stable barrier between the cells and the plasma.
These tubes are supplied for laboratory use only; they are labeled "not for human use."
Features
  • Ideal for use with the ABS210 (rev. 2019)
  • Simplified pipetting
  • Wide range of additives and fill volumes
Catalog No. NC2580246 Supplier Instech Supplier No. CTM500K3EDTA
Automated Blood Sampling Collection Tubes
The Microvette 300 and 500 tubes are ideal for use with the Instech Automated Blood Sampler due to the shape of the tube, which simplifies pipetting, and the wide range of additives and fill volumes.
The CT-M500-LIHEPG and CT-M500-SG models contain a unique inert acrylic gel at the bottom of the tube, under the additive, that moves upward during centrifugation to form a stable barrier between the cells and the plasma.
These tubes are supplied for laboratory use only; they are labeled "not for human use."
Features
  • Ideal for use with the ABS210 (rev. 2019)
  • Simplified pipetting
  • Wide range of additives and fill volumes