Learn More
Research Products International Corp Collection Tubes, 2ml, 1000 Pack
SDP
Catalog No. 501977881
Encompass
Durable polypropylene collection tube that is used in conjunction with the Zymo-Spin line of spin-columns (i.e., Zymo-Spin I through Zymo-Spin V). Capacity is 2 ml.
Catalog No. 50-197-7881 Supplier Research Products International Corp Supplier No. ZC10011000
Durable polypropylene collection tube that is used in conjunction with the Zymo-Spin line of spin-columns (i.e., Zymo-Spin I through Zymo-Spin V). Capacity is 2 ml.