Learn More
Perkin Elmer US LLC Collector Head Assembly for Auto-Ignite Wide Range FID
Catalog No. 502117286
Encompass_Preferred
Note that this collector assembly is not compatible with legacy Gas Chromatographs such as the Clarus 500/580/600/680.
Catalog No. 50-211-7286 Supplier Perkin Elmer US LLC Supplier No. N6550077
Note that this collector assembly is not compatible with legacy Gas Chromatographs such as the Clarus 500/580/600/680.