Learn More
Waters Corp Waters Collet and Compression Screw Multi-Tool, ACQUITY System, HFPT
Catalog No. NC9755301 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Collet and Compression Screw Multi-Tool is specifically engineered for use with ACQUITY UPLC systems, particularly with a Column Manager. This versatile tool functions primarily as a collet separator, facilitating the gentle disengagement of collets from gold compression screws within high-pressure finger-tight fittings.
  • Designed for use with ACQUITY UPLC systems.
  • Compatible with column manager components.
  • Facilitates separation of collets from gold compression screws.
  • Intended for high-pressure finger-tight fittings.
  • Packaged with one unit.
  • Replacement on an as-needed basis.
Catalog No. NC9755301 Supplier Waters Corp Supplier No. 700003170
The Waters Collet and Compression Screw Multi-Tool is specifically engineered for use with ACQUITY UPLC systems, particularly with a Column Manager. This versatile tool functions primarily as a collet separator, facilitating the gentle disengagement of collets from gold compression screws within high-pressure finger-tight fittings.
  • Designed for use with ACQUITY UPLC systems.
  • Compatible with column manager components.
  • Facilitates separation of collets from gold compression screws.
  • Intended for high-pressure finger-tight fittings.
  • Packaged with one unit.
  • Replacement on an as-needed basis.