Learn More
Waters Corp Waters Collet HPFT, Metal, ACQUITY UPLC Column Manager, 10 Pack
Catalog No. NC1854398 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Collet HPFT is a small metal ferrule designed for connecting a column to the preheater assembly of an ACQUITY UPLC Column Manager. It serves as a vital barrier between the outside environment and the connection, ensuring a solid link crucial for proper assay functionality. A worn-out collet can lead to connection failure, making it important to have replacements on hand.
  • Small metal ferrule
  • Designed for ACQUITY UPLC Column Manager
  • Sold in packs of 10
  • Ensures solid connection between column and preheater assembly
Catalog No. NC1854398 Supplier Waters Corp Supplier No. 700003168
The Collet HPFT is a small metal ferrule designed for connecting a column to the preheater assembly of an ACQUITY UPLC Column Manager. It serves as a vital barrier between the outside environment and the connection, ensuring a solid link crucial for proper assay functionality. A worn-out collet can lead to connection failure, making it important to have replacements on hand.
  • Small metal ferrule
  • Designed for ACQUITY UPLC Column Manager
  • Sold in packs of 10
  • Ensures solid connection between column and preheater assembly