Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNA Genotek™ Colli-Pee™ UAS First Void Urine Collection Device

Catalog No. 21279178
Change view
Click to view available options
Volume (Metric):
40 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-279-178 40 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-279-178 Supplier DNA Genotek™ Supplier No. N00342
Add to Cart
Add to Cart
This item is not returnable. View return policy

40 mL urine collection tube with UAS preservative, offers various sizes to capture a range of first-void urine volumes

  • Non-invasive, volumetric, standardized, self-collection of first-void urine samples
  • Offering various sizes to capture a range of first-void urine volumes, the Colli-Pee device is powering non-invasive self-sampling solutions with first-void urine.
  • Volumetric collection of first-void urine
  • UAS preservative
  • User preferred, easy self-collection
  • U.S. FDA Class 1 device
  • Device / Assay manufacturers must validate the use of this urine collection device for use with their device

Specifications

Product Type Urine Specimen Collection Kit
Closure Color Green
Closure Type Push-fit
Disposable Single-use
Graduated Yes
Material Polypropylene
Sterility Non-sterile
Color Clear
Diameter (Metric) Inner 27.3 mm
Includes UAS Preservative
Latex Free Yes
Volume (Metric) 40 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.