Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collibri™ DNA Library Preparation Kit Modules

Catalog No. A38579024
Encompass_Preferred
Change view
Click to view available options
No. of Samples:
24 Preps
96 Preps
Product Type:
DNA CD Indexes
DNA Library Cleanup Kit
DNA UD Indexes (Set A, 1-24)
DNA UD Indexes (Set B, 25-48)
DNA UD Indexes (Set C, 49-72)
DNA UD Indexes (Set D, 73-96)
ES DNA Library Prep
PCR-free ES DNA Library Prep
PCR-free PS DNA Library Prep
PS DNA Library Prep
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Samples Product Type
A38579024 24 Preps ES DNA Library Prep
A38575024 24 Preps PCR-free ES DNA Library Prep
A38577096 96 Preps PCR-free ES DNA Library Prep
A39097096 96 Preps ES DNA Library Prep
A38576024 24 Preps PCR-free PS DNA Library Prep
A39121096 96 Preps PCR-free PS DNA Library Prep
A39122024 24 Preps PS DNA Library Prep
A39099096 96 Preps PS DNA Library Prep
A38586024 24 Preps DNA CD Indexes
A38587024 24 Preps DNA UD Indexes (Set A, 1-24)
A43348024 24 Preps DNA UD Indexes (Set B, 25-48)
A43350024 24 Preps DNA UD Indexes (Set C, 49-72)
A43351024 24 Preps DNA UD Indexes (Set D, 73-96)
A43190024 24 Preps DNA Library Cleanup Kit
A38584096 96 Preps DNA Library Cleanup Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. A38579024 Supplier Invitrogen™ Supplier No. A38579024
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Invitrogen Collibri DNA Library Preparation Kit Modules are designed for a custom build of next-generation sequencing (NGS) workflow on Illumina systems.

The Invitrogen Collibri DNA Library Preparation Kit Modules are designed for a custom build of next-generation sequencing (NGS) workflow on Illumina systems. The kit supports whole-genome sequencing (WGS) applications and is suitable for all genomes. Modules are available in two sizes: for 24 or 96 samples.

The Invitrogen Collibri DNA Library Preparation Kit Modules are designed for a custom build of next-generation sequencing (NGS) workflow on Illumina systems. The kit supports whole-genome sequencing (WGS) applications and is suitable for all genomes.

The Collibri DNA Library Preparation Kit Modules support:

  • The need for enzymatic manipulation of DNA into a pool of cDNA fragments
  • The need to cleanup and size select library to get rid of junk sequences and purify a sample
  • The addition of indexed adapters to enable the prepared library sequencing on Illumina instruments

The enzymatic module of the kit generates a consistently high yield of a library, which is enough for library quantification, sequencing, and multiple runs on Illumina instruments.

Enzymatic Shearing (ES) modules

  • The Collibri ES DNA Library Preparation Kit Modules are designed for fast and convenient library preparation from a wide range of inputs (1–500 ng), starting from intact genomic DNA or already prepared amplicons (>300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow.
  • The Collibri PCR-free ES DNA Library Preparation Kit Modules are designed for fast and convenient library preparation from inputs as low as 100 ng intact genomic DNA or already prepared amplicons (>300 bp) of any species. The entire library preparation protocol takes about two hours for the PCR-free workflow.

Physical Shearing (PS) modules

  • The Collibri PS DNA Library Preparation Kit Modules offer library preparation of a wide range of inputs (1 ng to 1 μg) of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.
  • The Collibri PCR-free PS DNA Library Preparation Kit modules offer library preparation from inputs as low as 500 ng of physically sheared double-stranded DNA, cfDNA, or amplicons (≤300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 500 ng DNA.

If needed, Collibri Library Amplification Master Mix with primers can be bought separately. Or, choose the Collibri ES or PS DNA Library Preparation Kit Modules where these come included with the kit.

Collibri DNA Library Cleanup Kit contains exceptional beads optimized for work with DNA samples. Therefore, the DNA Library Preparation Kit works the best with these cleanup modules and is not recommended to be replaced with a regular bead.

Collibri DNA Indexed adapters contain the highest-quality Combinatorial Dual (CD), or Unique Dual (UD). For the higher throughput use UD and CD together to support up to 192 preparations per run. To avoid overlap of sequences, do not pool together different sizes of modules containing the same type of indexed adaptors (CDs or UDs only).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Description Fragmentation, End Repair/A-tailing step, Ligation, PCR
Format Kit
Sequencing Type Genome and DNA Sequencing
For Use With (Application) DNA Sequencing; Whole-Genome Sequencing, Amplicon Sequencing
No. of Samples 24 Preps
Product Line Collibri
Product Type ES DNA Library Prep
Quantity 24 preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.