Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Collibri™ ES DNA Library Prep Kits for Illumina Systems

Catalog No. A38607096W
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
24 Preps
96 Preps
Product Type:
ES DNA Library Prep Core Kit
ES DNA Library Prep Kit for Illumina Systems with CD Indexes
ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set A, 1-24)
ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set A-D, 1-96)
ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set B, 25-48)
ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set C, 49-72)
ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set D, 73-96)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Product Type
A38607096W 96 Preps ES DNA Library Prep Core Kit
A38605024 24 Preps ES DNA Library Prep Kit for Illumina Systems with CD Indexes
A38607096 96 Preps ES DNA Library Prep Kit for Illumina Systems with CD Indexes
A38606024 24 Preps ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set A, 1-24)
A43605024 24 Preps ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set B, 25-48)
A43606024 24 Preps ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set C, 49-72)
A43607024 24 Preps ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set D, 73-96)
A38607196 96 Preps ES DNA Library Prep Kit for Illumina Systems with UD Indexes (Set A-D, 1-96)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. A38607096W Supplier Invitrogen™ Supplier No. A38607096W
Add to Cart
Add to Cart

The Invitrogen Collibri ES DNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-genome sequencing (WGS) applications and is suitable for all genomes.

The Collibri ES DNA Library Preparation Kit is designed for fast and convenient library preparation from a wide range of inputs (1–500 ng), starting from intact genomic DNA or already prepared amplicons (>300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 100 ng DNA.

The Collibri ES DNA Library Preparation Kit is designed for fast and convenient library preparation from a wide range of inputs (1–500 ng), starting from intact genomic DNA or already prepared amplicons (>300 bp) of any species. The entire library preparation protocol takes about three hours with a PCR workflow. Additionally, a PCR-free version is available for inputs starting at 100 ng DNA.

The Collibri ES DNA Library Preparation Kit:
• Includes all necessary components in a single kit* for a quick use
• Is empowered on miniaturization liquid handlers to cut on costs
• Provides in-process visual feedback for easier handling
• Is suitable for conventional automation to increase throughput

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules, except for the Core kit, which enables Collibri combination with desired indexed adapters.

The Collibri ES DNA Library Preparation Kit contains inert dyes that provide visual feedback of preparation progress: the reaction mixes change color in response to the addition of critical components at each step. Moreover, the enzymatic shearing (ES) module of the kit generates a consistently high yield of library from any amount of input DNA (including the PCR-free version), which is enough for library quantification  before sequencing and multiple runs on Illumina instruments.

Applications
The Collibri ES DNA Library Preparation Kit is recommended for:
• Whole genome sequencing
• Resequencing
• De novo assembly

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Collibri ES DNA Library Prep Kit, store at -20°C
• Collibri DNA Library Cleanup Kit, store at 4°C
• Collibri DNA CD Indexes, store at -20°C
Description The product contains modules A39097096, A38584096
Sample Type DNA
Sequencing Type Genome and DNA Sequencing
For Use With (Equipment) HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NovaSeq 6000
For Use With (Application) Whole Genome Sequencing, Resequencing, De Novo Assembly
Libraries Fragment Library
No. of Reactions 96 Preps
Product Line Collibri
Product Type ES DNA Library Prep Core Kit
Quantity 96 preps
Shipping Condition Dry Ice
Workflow Step Library Generation
Show More Show Less
Are Collibri DNA Library Prep Core kits compatible with other commercially available indexed adaptors? I'm interested in performing higher throughput sequencing.

Collibri DNA Library Core kits are compatible with other commercially available set of adaptors such as Illumina TruSeq full-length adaptors, with eight base indexes.

Can indexed adaptors or other modules be purchased separately for the Collibri DNA Library Prep kits?

Indexed adapters and other modules in the kit can be purchased separately. Please refer to thermofisher.com/collibriDNA and contact us for more details.

With the Collibri DNA Library Prep kits, what is the distribution of enzymatically sheared DNA in the library?

Fragment size ranges from 150 bp to 850 bp. For optimal results, we recommend and provide detailed instructions for 350 bp and 550 bp average size library construction.

With the Collibri DNA Library Prep kits, can I use indexed adaptors interchangeably between RNA-seq and DNA-seq applications?

No. They are specifically designed for a product and cannot be used interchangeably.

With the Collibri DNA Library Prep kits, why is my ligation master mix orange instead of red?

Dyes inside the master mix react with CO2 and change the color from red to orange. The mix is still usable when the color changes to orange. To prevent discoloration, avoid keeping it on dry ice.

Are Collibri DNA Library Prep kits compatible with FFPE samples?

Collibri PS DNA Library Prep Kit can be used for library construction from lower-quality DNA (such as FFPE samples). However, the yields of adaptor-ligated molecules are typically lower compared to the yields obtained with high-quality DNA.

Collibri ES DNA Library Prep Kit is not recommended for library construction from FFPE samples.

How do I assess the quality of the prepared Collibri DNA library?

Library size distribution and the absence of primer dimers and/or over-amplification products should be verified by an electrophoretic method (with Agilent 2100 Bioanalyzer instrument).

To achieve the highest-quality sequencing data, it is essential to create optimal cluster densities across the flow cell. The most accurate quantification methods to assess the quantity of adaptor-ligated molecules in the library are qPCR-based methods, and we recommend using the Collibri Library Quantification Kit. Quantification can be performed at different stages of the workflow, for example after the post-ligation cleanup or size-selection step (for PCR-free workflows), or after the post-amplification cleanup step (for amplified libraries).

What is the PCR-free limit for Collibri PS and ES DNA Library prep kits?

PCR-free libraries of ≥4 nM concentration using Collibri PS DNA Library Preparation Kit can be prepared from as little as 500 ng input DNA.

PCR-free libraries of ≥4 nM concentration using Collibri ES DNA Library Preparation Kit can be prepared from as little as 100 ng input DNA.

In the Collibri DNA Library Prep kits, what enzyme is used for library amplification?

Collibri kits contain Platinum SuperFi DNA Polymerase whose exceptionally strong proofreading activity helps ensure amplification of NGS libraries with supreme sequence accuracy, producing NGS error rates similar to PCR-free methods. The enzyme is a part of Collibri Library Amplification Master Mix, which is included in the kits with PCR option. Designed exclusively for NGS application, Collibri Library Amplification Master Mix is based on a proprietary reaction buffer that has been specially optimized for efficient and uniform amplification of NGS libraries regardless of GC content.

Note: Collibri DNA Library Prep kits are available with or without PCR library amplification.

Are Collibri DNA Library Prep reagents available in a customized, larger format?

Yes, customized formats can be accommodated by our custom services group. Please send an email to techsupport@thermofisher.com with your custom request and they will provide you with further details.

What do ES and PS abbreviations stand for in the Collibri DNA Library Prep kits?

ES - Enzymatically sheared. Collibri ES DNA Library Prep Kit includes reagents for enzymatic shearing and downstream DNA library preparation.
PS - Physically sheared. Collibri PS DNA Library Prep Kit is designed for library construction starting from physically sheared DNA.

Are adaptors included in the Collibri DNA Library Prep kits?

Yes, adaptors are included in the Collibri DNA Library Prep kits except in DNA Library Prep Core kits that contain only enzymatic and clean-up modules. The Collibri DNA Library Prep kits include ready-to-ligate adaptors with unique dual indexes (UDs), or combinatorial dual indexes (CDs) that allow pooling of up to 24 or 96 different samples with one kit before the sequencing run.

Each well in the combinatorial dual indexes (CDs) plate contains a single-use adaptor that consists of a unique combination of two 8 nucleotide identification barcodes. Combination of one D5 barcode with one D7 barcode in each ready-to-use adaptor allows pooling of up to 24 or 96 different samples before the sequencing run.

Each well in the unique dual indexes (UDs) plate contains a single-use adaptor that consists of two unique 8-nucleotide indexes of D5 and D7 when each index appears in the whole plate only once. The combinatorial approach is not used.

Use CDs and UDs to pool up to 192 preps for higher throughput. Other combinations are also possible, but do not pool together different sizes of kits containing the same type of indexed adaptors (CDs or UDs only) to avoid overlap of sequences.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.