Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Collision in Two Dimensional
GREENER_CHOICESCI_ED

Catalog No. S139425 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139425 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139425 Supplier Supertek™ Supplier No. PH10517
Add to Cart
Add to Cart
This item is not returnable. View return policy

This Supertek™ Collsion in Two Demensional apparatus uses a curved ramp to deliver a ball for collision with constant energy.

  • Kit includes:
  • 1- Curved ramp
  • 2 - 1/2 in. steel balls
  • 1 - 1/2 in. glass ball
  • 1 - 1 in. wooden plump bob
  • 1 - String
  • 1 - Eyelet
  • Instructions included

Specifications

Grade 6+
Format Kit
Languages English
Subject Physics and Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.