Learn More
GSC Go Science Crazy Collisions in Two Dimensions Demonstration
SCI_ED
Catalog No. S13635 Shop All GSC Products
Click to view available options
For Use With (Application):
Demonstrating laws of conservation of momentum and energy
Catalog No. For Use With (Application)
S13635 Demonstrating laws of conservation of momentum and energy
1 options
Intended for ages 13 and up.
Catalog No. S13635 Supplier GSC Go Science Crazy Supplier No. 462412
Intended for ages 13 and up.
Includes
2 metal balls; 1 wooden ball; 1 glass ball; String; Metal apparatus; Instructions
Demonstrate the laws of conservation of momentum and energy. The GSC International Collisions in Two Dimensions Demonstration tests what happens when two objects collide and move off in different directions. This demonstration is intended for classroom applications.
  • Students to explore the laws of momentum and energy
  • Students test what happens when two objects collide and move off in different directions
  • Demonstration includes two metal balls, a wooden ball, a glass ball, string, and instructions
  • Apparatus easily clamps to a surface for stability
Specifications
For Use With (Application) Demonstrating laws of conservation of momentum and energy
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov