Learn More
Electron Microscopy Sciences Collodion, 5% Solution, USP 100 ML
SDP
Catalog No. 50980304
Encompass_Preferred
A solution of 25% Alcohol and 75% Diethyl Ether in Pyroxylin. Offered in a 30 ml bottle to avoid its classification as a hazardous chemical for shipping purposes.
Catalog No. 50-980-304 Supplier Electron Microscopy Sciences Supplier No. 12600
A solution of 25% Alcohol and 75% Diethyl Ether in Pyroxylin. Offered in a 30 ml bottle to avoid its classification as a hazardous chemical for shipping purposes.