Learn More
Electron Microscopy Sciences Collodion (Parlodion), 2% in Amyl Acetate 30 ML
SDP
Catalog No. 50980308
Encompass_Preferred
Highly purified nitrocellulose (parlodion strip) in glass distilled amyl acetate. Useful for forming a negative replica to very fine detail. Two types are available: Our sterile formula which is filtered down to 0.45 micron and our non-sterile formula. Non-Filter 2% Collodion. 30 ml container avoids hazardous shipping.
Catalog No. 50-980-308 Supplier Electron Microscopy Sciences Supplier No. 1262030
Highly purified nitrocellulose (parlodion strip) in glass distilled amyl acetate. Useful for forming a negative replica to very fine detail. Two types are available: Our sterile formula which is filtered down to 0.45 micron and our non-sterile formula. Non-Filter 2% Collodion. 30 ml container avoids hazardous shipping.