Learn More
Electron Microscopy Sciences Collodion (Parlodion), in Amyl Acetate 30 ML
SDP
Catalog No. 50980311
Encompass_Preferred
Highly purified nitrocellulose (parlodion strip) in glass distilled amyl acetate. Useful for forming a negative replica to very fine detail. Two types are available: Our sterile formula which is filtered down to 0.45 micron and our non-sterile formula. Sterile formula is filtered to 0.45 micron. 30 ml avoids hazardous shipping.
Catalog No. 50-980-311 Supplier Electron Microscopy Sciences Supplier No. 1262055
Highly purified nitrocellulose (parlodion strip) in glass distilled amyl acetate. Useful for forming a negative replica to very fine detail. Two types are available: Our sterile formula which is filtered down to 0.45 micron and our non-sterile formula. Sterile formula is filtered to 0.45 micron. 30 ml avoids hazardous shipping.