Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Collodion solution

Catalog No. 0998625L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-986-25L 2.5 L
09-986-500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-986-25L Supplier Sigma Aldrich Supplier No. 099862.5L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Collodion solution is used:
  • In the nitrocellulose treatment of working electrode in the tissue implanted biological oxygen sensor to prevent electrode poisoning
  • As a stabilizer in synthesizing silver nanoparticles (agnps) to prepare silver-based conductive printing ink
  • As a binder in the preparation of titanium powder by metallothermic reduction method

Specifications

CAS 9004-70-0
MDL Number MFCD00081525
Quantity 2.5 L
Synonym Cellulose nitrate
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.