Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collodion, Thermo Scientific Chemicals

Catalog No. AC405355000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405355000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC405355000 Supplier Thermo Scientific Chemicals Supplier No. 405355000

missing translation for 'validMainframeMsg'

CAS: 9004-70-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Collodion
Color Colorless to Yellow
Physical Form Viscous Liquid
CAS 9004-70-0, 60-29-7, 64-17-5, 8001-79-4
MDL Number MFCD00081525
Density (25/4°C) 0.775 to 0.79g/mL
Merck Index 15, 2468
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.