Learn More
DIAGNOSTIC CONSULTING NETWORK LLC COLLOIDAL GOLD 1L
SDP
Supplier:  DIAGNOSTIC CONSULTING NETWORK LLC CG0101L
Encompass
DCNovations Colloidal Gold has been optimized specifically for use in lateral flow applications. The suspension is manufactured at OD 1 with the average particle size around 25nm +/- 5nm. However, there is a wide size distribution, which DCN Dx scientists have found to improve assay sensitivity and conjugate stability. 1L; DCNOVATIONS mfg#CG-010(1L)
Catalog No. NC1949696