Learn More
Electron Microscopy Sciences EMS Colloidal Graphite, Alcohol Base 30 GR
SDP
Catalog No. 50980322
Encompass_Preferred
Isopropal base collodal graphite has a Flat surface texture. The average flake size is 1 micron. Service temperature is 204 degree C.
Catalog No. 50-980-322 Supplier Electron Microscopy Sciences Supplier No. 12660
Isopropal base collodal graphite has a Flat surface texture. The average flake size is 1 micron. Service temperature is 204 degree C.