Learn More
Electron Microscopy Sciences EMS Colloidal Graphite, Water Base 50 GR
SDP
Catalog No. 50980321
Encompass_Preferred
Water based collodal graphite has a flat surface texture. The average flake size is 1 um. Service temperature is 306 deg C.
Catalog No. 50-980-321 Supplier Electron Microscopy Sciences Supplier No. 12650
Water based collodal graphite has a flat surface texture. The average flake size is 1 um. Service temperature is 306 deg C.