Learn More
Diagnostic Biosystems COLLOIDAL IRON Colloidal Iron
Catalog No. 502407342
Encompass_Preferred
The Colloidal Iron Stain Kit is designed for the histological visualization of acid mucopolysaccharides.
Catalog No. 50-240-7342 Supplier Diagnostic Biosystems Supplier No. KT007
The Colloidal Iron Stain Kit is designed for the histological visualization of acid mucopolysaccharides.