Learn More
StatLab Colloidal Iron MasterTech Special Stain Kit
SDP
Catalog No. 22000050
Change view
Click to view available options
100 mL
Catalog No.
22-000-050 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-050 Supplier StatLab Supplier No. KTCIR
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains for acid mucosubstances in just 45 minutes
  • Working Colloidal Iron Solution is absorbed and reacts with a working Prussian Blue Solution, serving as the primary stain by producing blue acid mucosubstances against pink to red collagen counterstained by Van Gieson's
  • 100 mL Stain Kit yields between 144 and 225 slides
  • A MasterTech Special Stain Kit
Specifications
Product Type Iron Stain Kit