Learn More
Electron Microscopy Sciences Colloidal Silica, 1 Pint 16 OZ
SDP
Catalog No. 50997309
Encompass_Preferred
This non-crystallizing Colloidal Silica is made to be user friendly. Eliminates the problems that are caused by drying or freezing that are associated with other colloidal silica products which are used for chemical/mechanical polishing.
Colloidal silica is a first choice silica dispersion for optimizing polishing results such as silicon, fused quartz, fused silica, lithium niobate, YAZ, GGG, alexandrite, sapphire and many others.
Can be used full strength or diluted. Dilution to 1:1 or higher with deionized water is recommended. Polyurethane and short nap cloths can be used.
Catalog No. 50-997-309 Supplier Electron Microscopy Sciences Supplier No. 5036605
This non-crystallizing Colloidal Silica is made to be user friendly. Eliminates the problems that are caused by drying or freezing that are associated with other colloidal silica products which are used for chemical/mechanical polishing.
Colloidal silica is a first choice silica dispersion for optimizing polishing results such as silicon, fused quartz, fused silica, lithium niobate, YAZ, GGG, alexandrite, sapphire and many others.
Can be used full strength or diluted. Dilution to 1:1 or higher with deionized water is recommended. Polyurethane and short nap cloths can be used.