Learn More
Ted Pella Inc PELCO Colloidal Silver, 15g
SDP
Catalog No. NC9763514
Encompass
Air drying silver paint to make effective ground. Curable at room temperature and has high adhesion to any material. Surfaces do not have to be prepared prior to application and will adhere to such materials as polymer (phenolic) boards, ceramic, glass, metal, plastic and fiberglass. It should be mixed well with a spatula before using. RoHS compliant.
Catalog No. NC9763514 Supplier Ted Pella Inc Supplier No. 16034
Air drying silver paint to make effective ground. Curable at room temperature and has high adhesion to any material. Surfaces do not have to be prepared prior to application and will adhere to such materials as polymer (phenolic) boards, ceramic, glass, metal, plastic and fiberglass. It should be mixed well with a spatula before using. RoHS compliant.