Learn More
Electron Microscopy Sciences Colloidal Silver Liquid, 15 g
SDP
Catalog No. 50980312
Encompass_Preferred
1-Methoxy-2-propanol base. Fast drying. Average grain size less than 1 µm and Silver content is 60%. Sheet resistance is 0.02 ohm per square inch at 1 mil thickness. Comes with a brush attached to the cap. Service temperature is 30 minutes at 200°C.
Catalog No. 50-980-312 Supplier Electron Microscopy Sciences Supplier No. 12630
1-Methoxy-2-propanol base. Fast drying. Average grain size less than 1 µm and Silver content is 60%. Sheet resistance is 0.02 ohm per square inch at 1 mil thickness. Comes with a brush attached to the cap. Service temperature is 30 minutes at 200°C.