Learn More
Ted Pella Inc PELCO Colloidal Silver Paste, 25g
SDP
Catalog No. NC9707737
Encompass
Ideal where a nonflowing paste is needed for special applications. Particle size: 0.4-1.0µm, 80% are below 1.0µm. Cures in 16-20 hours, or 30 minutes @125-150°C. Clear lacquer base. Approximate Specific Gravity 2.25g/cc3. Service temperature = 200°C. Use #16021 gold/silver thinner/extender to dilute if needed.
Catalog No. NC9707737 Supplier Ted Pella Inc Supplier No. 16032
Ideal where a nonflowing paste is needed for special applications. Particle size: 0.4-1.0µm, 80% are below 1.0µm. Cures in 16-20 hours, or 30 minutes @125-150°C. Clear lacquer base. Approximate Specific Gravity 2.25g/cc3. Service temperature = 200°C. Use #16021 gold/silver thinner/extender to dilute if needed.