Learn More
Electron Microscopy Sciences Silver Paste, 25 g
SDP
Catalog No. 50980313
Encompass_Preferred
Thick base, ideal for non-flowing required. Easily applied with micro spatula or wooden toothpick. Particle size ranging 0.4-1 m, 80% are less than 1 m. Cure in 16 to 20 hours at room temperature or 30 minutes at 125-150 C.
Catalog No. 50-980-313 Supplier Electron Microscopy Sciences Supplier No. 12640
Thick base, ideal for non-flowing required. Easily applied with micro spatula or wooden toothpick. Particle size ranging 0.4-1 m, 80% are less than 1 m. Cure in 16 to 20 hours at room temperature or 30 minutes at 125-150 C.