Learn More
Newcomer Supply Colloidal Iron Control Slides, 10/set
SDP
Catalog No. NC0712842
Encompass
Newcomer Supply Colloidal Iron Control Slides are for the positive histochemical staining of acid epithelial mucins (sialomucin, sulfomucin) and stromal (mesenchymal) mucin in tissue sections. Colloidal Iron Control Slides contain sections of positive staining small intestine.
Catalog No. NC0712842 Supplier Newcomer Supply Supplier No. 4127A
Newcomer Supply Colloidal Iron Control Slides are for the positive histochemical staining of acid epithelial mucins (sialomucin, sulfomucin) and stromal (mesenchymal) mucin in tissue sections. Colloidal Iron Control Slides contain sections of positive staining small intestine.