Learn More
LAMDA BIOTECH INC Column-Pure™ Bacterial Genomic DNA Kit
Catalog No. 502045802
Encompass_Preferred
This is a quick and easy spin column method designed for rapid isolation of genomic DNA from cells and bacteria. The kit contains a membrane embedded column for binding up to 10 μg of genomic DNA. Nucleotides, proteins, salts, and other impurities are washed away. Purified genomic DNA can be used in most molecular biology experiments including restriction enzyme digestion, PCR, Southern-blotting, etc.
Catalog No. 50-204-5802 Supplier LAMDA BIOTECH INC Supplier No. D423100
This is a quick and easy spin column method designed for rapid isolation of genomic DNA from cells and bacteria. The kit contains a membrane embedded column for binding up to 10 μg of genomic DNA. Nucleotides, proteins, salts, and other impurities are washed away. Purified genomic DNA can be used in most molecular biology experiments including restriction enzyme digestion, PCR, Southern-blotting, etc.